Photometric tube tests, NANOCOLOR® Manganese
Supplier: Macherey-Nagel
NANOCOLOR®
MANA985058EA 87.6 EUR
MANA985058
Photometric tube tests, NANOCOLOR® Manganese
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of manganese in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.