(R)-3-(2-Bromophenyl)piperidine hydrochloride
Supplier: Apollo Scientific
OR321522-1G OR321522-250MG
APOSOR321522-1GEA 1120 EUR
APOSOR321522-1G APOSOR321522-250MG
(R)-3-(2-Bromophenyl)piperidine hydrochloride
(R)-3-(2-Bromophenyl)piperidine hydrochloride
Formula: C₁₁H₁₄BrN.ClH
MW: 276.601 g/mol
Storage Temperature: Refrigerator
CAS Number: 1336565-35-5