1-(1-Cyclopropyl-1H-pyrazol-4-yl)methanamine dihydrochloride 95%
Supplier: Apollo Scientific
Warning
OR475192-1G OR475192-250MG
APOSOR475192-1GEA 719 EUR
APOSOR475192-1G APOSOR475192-250MG
1-(1-Cyclopropyl-1H-pyrazol-4-yl)methanamine dihydrochloride 95%
1-(1-Cyclopropyl-1H-pyrazol-4-yl)methanamine dihydrochloride
Formula: C₇H₁₁N₃.₂ClH
MW: 210.104 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32669858
CAS Number: 2514952-94-2