[(4-Bromo-1-cyclopropyl-1H-pyrazol-3-yl)methyl](methyl)amine dihydrochloride 95%
Supplier: Apollo Scientific
Warning
OR475197-1G OR475197-250MG
APOSOR475197-1GEA 719 EUR
APOSOR475197-1G APOSOR475197-250MG
[(4-Bromo-1-cyclopropyl-1H-pyrazol-3-yl)methyl](methyl)amine dihydrochloride 95%
[(4-Bromo-1-cyclopropyl-1H-pyrazol-3-yl)methyl](methyl)amine dihydrochloride
Formula: C₈H₁₂BrN₃.₂ClH
MW: 303.027 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32663471
CAS Number: 2514953-00-3