3-Bromo-5,6-difluoro-2-methoxybenzamide
Supplier: Apollo Scientific
PC303495-1G PC303495-5G
APOSPC303495-1GEA 279 EUR
APOSPC303495-1G APOSPC303495-5G
3-Bromo-5,6-difluoro-2-methoxybenzamide
3-Bromo-5,6-difluoro-2-methoxybenzamide
Formula: C₈H₆BrF₂NO₂
MW: 266.04 g/mol
Storage Temperature: Ambient
CAS Number: 2512221-83-7