2-[2-Methyl-3-(trifluoromethyl)phenyl]propan-2-ol ≥98%
Supplier: Apollo Scientific
PC52478-10G PC52478-1G PC52478-5G
APOSPC52478-10GEA 574 EUR
APOSPC52478-10G APOSPC52478-1G APOSPC52478-5G
2-[2-Methyl-3-(trifluoromethyl)phenyl]propan-2-ol ≥98%
2-[2-Methyl-3-(trifluoromethyl)phenyl]propan-2-ol
Formula: C₁₁H₁₃F₃O
MW: 218.216 g/mol
Storage Temperature: Ambient
CAS Number: 2514941-96-7