Human LHRH
Supplier: Apollo Scientific
BITH3030-100MG BITH3030-10MG BITH3030-25MG
APOSBITH3030-100MGEA 0 EUR
APOSBITH3030-100MG APOSBITH3030-10MG APOSBITH3030-25MG
Human LHRH
Proteins and Peptides
Order Now
SPECIFICATIONS
Protein/peptide name LHRH
Environmentally Preferable
Protein synonyms Leutenizing hormone Releasing Hormone (Gonadorelin)
Species Human
Source
Learn more