NANOCOLOR® Photometric Tube Tests, Total Nitrogen
Supplier: MACHEREY-NAGEL
985088 985083 985092
MANA985088EA 113 EUR
MANA985088 MANA985083 MANA985092
NANOCOLOR® Photometric Tube Tests, Total Nitrogen
Environmental Test Kits
Photometric test kits for the determination of total nitrogen in water samples.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Colored pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (AD) round cuvettes. They are pre-programmed on NANOCOLOR® photometers and are also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: No
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent: No
Digestion: No
NanOx metal: No
NanOx: Yes
Digestion temperature: 120 °C
Delivery information:
2 rugged boxes with 20 test tubes for decomposition and 20 test tubes for analysis. Sufficient for 20 tests.