Enzyme test pipette, conformity certified, Assistent
Supplier: Hecht
HECH40545724EA 89.5 EUR
HECH40545724
Enzyme test pipette, conformity certified, Assistent
Pipettes Haemometer Pipettes
Clear glass, with elongated strong tip, precision type, for partial delivery (not calibrated to tip).
  • Calibrated to deliver (Ex), short delivery time
  • Zero point at top, main points
  • Fused amber graduation 'ASSISTENT-PERMANENT'
Available with individual certificate on request (extra charge).