Tryptone
Supplier: Apollo Scientific
APOSBIT1332-1KGEA 256 EUR
APOSBIT1332-1KG APOSBIT1332-500G
Tryptone
Media for Microbiology Dehydrated Media for Microbiology
Pancreatic digest of caseinTypical analysis (% w/w) total nitrogen: approx. 12,7, amino nitrogen: approx. 3,7, an:tn ratio: approx. 0,26 sodium chloride: approx. 0,4, tryptophan: approx. 1,0 pH(1% solution) 7,3.
  • MFCD00131829