Capmats for J.T.Baker® Plate Round Well
Supplier: Ritter, part of Avantor
J.T.Baker®
43001-0017EA 29.75 EUR
43001-0017
Capmats for J.T.Baker® Plate Round Well
Seals Microplate Seals
Suitable to use with J.T.Baker® Plate Round Well 43001-0010, 43001-0016 or 43001-0116.