HT (high temperature) laboratory connectors with GL thread
Supplier: Bohlender
BOLA
201-1014EA 80.6 EUR
201-1014 201-1015 201-0960 201-0961 201-1016 201-1017 201-0962 201-1011 201-1012 201-1013 201-0956 BOHLD641-84 201-0957 201-0958 201-0959 201-0952 201-0953 201-0954 201-0955 201-1928 201-1929 201-1020 201-1267 201-0970 201-1025 201-1026 201-1268 201-0971 201-1027 201-1269 201-0972 201-0973 201-1028 201-1021 201-1022 201-1264 201-1023 201-1265 201-1266 201-1024 201-0967 201-0968 201-0969 201-0963 201-1018 201-0964 201-1019 201-0965 201-0966 201-0950 201-0951 201-1002 201-0949 201-1292 201-1290 201-1291 229-0089 229-0088 201-0996 201-0997 201-0998 201-0999 229-0094 229-0093 229-0092 229-0091 229-0090 229-0099 229-0098 229-0097 229-0096 229-0095 201-1270 201-1271 201-1272 201-1030 201-1273 201-0981 201-1278 201-1279 201-0982 201-0983 201-0984 201-1274 201-1275 201-1276 201-1277 201-0980 229-0102 201-0978 229-0101 201-0979 229-0100 201-1029 201-0974 201-0975 201-0976 201-0977 229-0109 229-0108 229-0105 229-0104 229-0103 201-1281 201-1282 201-1283 201-1284 201-1280 201-1289 201-0992 201-0993 201-0994 201-0995 201-1285 201-1286 201-0990 201-1287 201-1288 201-0991 201-0989 201-0985 201-0986 201-0987 201-0988
HT (high temperature) laboratory connectors with GL thread
Fittings and Connectors
Screw cap and clamping ring made from PPS, sealing ring and tapered ring (ferrule) made from PTFE.
  • Good chemical resistance
  • Temperature resistant from –50 to +250 °C
  • Pressure resistant up to a maximum of 10 bar, vacuum suitable
To connect apparatus and fittings with GL threads and hard-walled hoses or tubes made from glass, plastic or metal. To lock probes, thermometers, immersion tubes or cable feeds inserted in reaction vessels. Additional o-ring on screw connections for hose external diameters of less than 3 mm (not exposed to nor in contact with the fluid medium).
  • Good chemical resistance
  • Temperature resistant from –50 to +250 °C
  • Pressure resistant up to a maximum of 10 bar, vacuum suitable
To connect apparatus and fittings with GL threads and hard-walled hoses or tubes made from glass, plastic or metal. To lock probes, thermometers, immersion tubes or cable feeds inserted in reaction vessels. Additional o-ring on screw connections for hose external diameters of less than 3 mm (not exposed to nor in contact with the fluid medium).