CryoBoxes, Cardboard, Origami
Supplier: TENAK
TE22825 TE22830
479-1175EA 223 EUR
479-1175 479-1176
CryoBoxes, Cardboard, Origami
Boxes Cryoboxes
Cryoboxes, white cardboard, PP coated.
  • PP coated for longer durability
  • Easy to assemble
  • Water-repellant
  • Delivered 'flat-packed' and unassembled: Saves shipping cost and storage space in the laboratory