Capsaicin
Supplier: Apollo Scientific
Synonyms: 8-Methyl-N-vanillyl-6-none-amid
BIC0125-5G BIC0125-10G BIC0125-25G
APOSBIC0125-5GEA 132 EUR
APOSBIC0125-5G APOSBIC0125-10G APOSBIC0125-25G
Capsaicin
Capsaicin
Active component of cayenne pepper.
Formula: C₁₈H₂₇NO₃
Boiling Pt: 210…220 °C
Melting Pt: 65…68 °C
Flash Pt: 113 °C
MDL Number: MFCD00017259
CAS Number: 404-86-4