Puromycin dihydrochloride, powder
Supplier: Corning
MDTC61-385-RAEA 243.51 EUR
MDTC61-385-RA
Puromycin dihydrochloride, powder
Puromycin dihydrochloride
Puromycin dihydrochloride is a nucleoside antibiotic and protein synthesis inhibitor.
By acting as an analog of the 3'-terminal end of aminoacyl-tRNA, it prevents growth of bacteria, protozoa, algae, and mammalian cells by causing premature protein termination.
The resistance gene (puromycin acetyltransferase) gives very effective protection. It is effective against Gram-negative and Gram-positive bacteria.
Balení: Shipped frozen.
Formula: C₂₂H₂₉N₇O₅·2HCl
MW: 544,44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012691
CAS Number: 58-58-2