Diethylenetriaminepentaacetic acid ≥99%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Warning

85989.180EA 97.2 EUR
85989.180
Diethylenetriaminepentaacetic acid ≥99%, AnalaR NORMAPUR® analytical reagent
Diethylenetriaminepentaacetic acid
Formula: C₁₄H₂₃N₃O₁₀
MW: 393,35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004289
EINECS: 200-652-8
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay (dried basis) ≥ 99 %
Identification (IR) Passes test
pH (saturated water) 2.0 - 3.0
Heavy metals (as Pb) ≤ 10 ppm
Ignition residue (SO4) ≤ 0.15 %
Water ≤ 0.5 %
Ba (Barium) ≤ 5 ppm
Ca (Calcium) ≤ 10 ppm
Cd (Cadmium) ≤ 5 ppm
Co (Cobalt) ≤ 5 ppm
Cr (Chromium) ≤ 5 ppm
Cu (Copper) ≤ 5 ppm
Fe (Iron) ≤ 10 ppm
K (Potassium) ≤ 50 ppm
Mg (Magnesium) ≤ 100 ppm
Mn (Manganese) ≤ 5 ppm
Na (Sodium) ≤ 0.15 %
Ni (Nickel) ≤ 5 ppm
Zn (Zinc) ≤ 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR