Grade 520 bII ½ filter papers for technical use, Whatman™
Supplier: Whatman products (Cytiva)
10331551 10331553 10331554 10331556 10331558
515-8030EA 113 EUR
515-8030 WHAT10331553 515-8064 515-8036 515-8038
Grade 520 bII ½ filter papers for technical use, Whatman™
Filters Filter Papers Qualitative Filter Paper
Grade 520bll ½ thick, pleated filter paper with high wet strength offering a very high flow rate.
Order Now
SPECIFICATIONS
Grade 520 bII ½
Environmentally Preferable
Weight 155 g/m²
Thickness 550 µm
Particle retention 20 µm
Filtration speed 30 (Herzberg)