Ampicillin sodium salt, powder
Supplier: Corning
Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt
61-238-RM 61-238-RH
MDTC61-238-RMEA 367.9 EUR
MDTC61-238-RM MDTC61-238-RH
Ampicillin sodium salt, powder
Ampicillin sodium salt
Ampicillin is a Penicillin derivative that inhibits bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane.
A ß-lactam antibiotic with an amino group side chain attached to the penicillin structure, its mode of resistance is through the cleavage of ß-lactam ring of ampicillin by ß-lactamase.
Ampicillin is effective against Gram-positive and some Gram-negative bacteria.
Balení: Shipping temperature: Ambient
Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3