Ciprofloxacin hydrochloride, powder
Supplier: Corning
Synonyms: 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, 1-Cyclopropyl- 6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid hydrochloride
61-277-RF 61-277-RG
MDTC61-277-RFEA 184.48 EUR
MDTC61-277-RF MDTC61-277-RG
Ciprofloxacin hydrochloride, powder
Ciprofloxacin hydrochloride
Ciprofloxacin Hydrochloride (HCl) is a 4-fluoroquinolone antibiotic often used to control mycoplasma contamination in infected cell lines
It works by interfering with the bacterial enzyme DNA gyrase, which is necessary for bacterial synthesis, replication, and transcription in both the active and non-active growth phases of the bacterial life cycle. Ciprofloxacin HCl is effective against a wide range of Gram-positive and Gram-negative bacteria and is offered in powder form.
Formula: C₁₇H₁₈FN₃O₃·HCl
MW: 367.84 g/mol
MDL Number: MFCD00079044
CAS Number: 86393-32-0