High-Flow Nylon Monofilament Filter Bags
Supplier: Pentair
MFLX01524-58EA 7.1 EUR
MFLX01524-58
High-Flow Nylon Monofilament Filter Bags
Filters Bag Filter Systems
Wide range of options for temperature, flow rate, and filtration quality.
  • Constructed of nylon
Designed for higher flow with an average efficiency at the rated pore size. The bags are glazed to minimize fiber migration. Nylon monofilament is a highstrength bag for handling a relatively more aggressive, cruder filtration application. These have a temperature rating of 300 °F (149 °C) maximum.