Extra-Strength Pleated Cellulose Cartridges
Supplier: Avantor Fluid Handling
MFLX01509-11EA 14 EUR
MFLX01509-11 MFLX01509-09
Extra-Strength Pleated Cellulose Cartridges
Filters Filter Cartridges
Pleated filter gives you more surface area for higher flowrate and longer life.
  • Constructed of cellulose and polyester
This cellulose cartridge is blended with polyester for better wet strength. The polyester material allows more pleats to improve flow and dirt loading. Suitable to 165 °F (74 °C).
Note: For optimal performance, change cartridge at 25 psid.