Reducer Fittings, Male NPT Threaded, Straight
Supplier: Avantor Fluid Handling
MFLX06546-30EA 37.3 EUR
MFLX06546-30 MFLX06546-23 MFLX06546-27
Reducer Fittings, Male NPT Threaded, Straight
Fittings and Connectors
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.