High Capacity Battery TR-632, Versaflo™
Supplier: 3M
Versaflo™
MMMATR-632EA 757 EUR
MMMATR-632
High Capacity Battery TR-632, Versaflo™
Batteries
The Versaflo™ High Capacity Battery TR-632 is a rechargeable lithium-ion battery designed for use with the Versaflo™ Powered Air Turbo TR-600 Series.
  • Lightweight, high-capacity, lithium-ion battery pack
  • Charges in under four hours with light-up charge level indicator
  • Provides extended duration when compared to TR-630 standard battery
  • Rinseable and temporarily submergible for easy cleaning
  • Easy to see battery charge level
  • CE approved
A new high capacity battery can provide a turbo system with 9,5 to 19 hours of use at a standard flow rate when equipped with a new particulate filter (TR-6710E). System duration will vary based on battery age, filter loading, filter selection, and user selected airflow setting. Under equivalent conditions, the TR-632 high capacity battery provides extended system durations when compared to the TR-630 standard battery.
This battery back charges in less than four hours and features a light-up charge level indicator. It's practical and can be rinsed or temporarily submerged in water for easy cleaning. See User Instructions for further information.