Battery charger kit TR-641E, Versaflo™
Supplier: 3M
Versaflo™
111-2549EA 434 EUR
111-2549
Battery charger kit TR-641E, Versaflo™
Batteries
The Versaflo™ Battery Charger Kit TR-641E is a single-station battery charger which consists of a TR-640 charging cradle, power supply, and lead. It's designed for use with all batteries from the Versaflo™ Powered Air Turbo TR-600 Series.
  • Charges in under four hours
  • Charges one battery at a time
  • Store fully-charged batteries on charger, ready for immediate use
  • CE approved
This kit charges the Versaflo™ standard TR-630 battery or the Versaflo™ high-capacity TR-632 battery individually in less than four hours. The battery can then be stored in the cradle once fully charged, ready for immediate use. The kit comes with a charging cradle, power supply, and lead.