Ammonium iron(III) sulphate dodecahydrate ACS
Supplier: Merck
Warning
SIAL221260-500GEA 67.2 EUR
SIAL221260-500G
Ammonium iron(III) sulphate dodecahydrate ACS
Ammonium iron(III) sulphate dodecahydrate
Formula: NH₄Fe(SO₄)₂·12H₂O
MW: 482,2 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 39…41 °C
Density: 1,71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150004
CAS Number: 7783-83-7
EINECS: 233-382-4
Merck Index: 12,00549