n-Decane ≥99%, ReagentPlus®
Supplier: Merck
Danger
D901-100ML D901-500ML
SIALD901-100MLEA 57.3 EUR
SIALD901-100ML SIALD901-500ML
n-Decane ≥99%, ReagentPlus®
n-Decane
Formula: H₃C(CH₂)₈CH₃
Boiling Pt: 174.1 °C (1013 hPa)
Melting Pt: –30 °C
Density: 0.73 g/cm³ (20 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00008954
CAS Number: 124-18-5
EINECS: 204-686-4
UN: 2247
ADR: 3,III