Potassium carbonate, anhydrous 99 zS
Supplier: VWR Chemicals
Warning
Synonyms: Potash
91239.320EA 101 EUR
91239.320 91239.230
Potassium carbonate, anhydrous 99 zS
Potassium carbonate
Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891 °C
Density: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
Order Now
Specification Test Results
Assay Min. 99 %