n-Nonane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Danger
87100.180EA 24.45 EUR
87100.180 87100.290 87100.320 87100.230
n-Nonane ≥99%, GPR RECTAPUR®
n-Nonane
Formula: H₃C(CH₂)₇CH₃
MW: 128,26 g/mol
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00009574
EINECS: 203-913-4
UN: 1920
ADR: 3,III
Order Now
Specification Test Results
Assay ≥ 99,0 %
Density (20/4) 0,717 - 0,719