Sorafenib tosylate (BAY 43-9006 tosylate salt)
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide, 4-methylbenzenesulphonate, Sorafenib Tosylate, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N′-[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea, 4-methylbenzenesulphonate, 4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)phenoxy)-N-methylpicolinamide 4-methylbenzenesulphonate
465710010 465712500
ACRO465710010EA 259 EUR
ACRO465710010 ACRO465712500
Sorafenib tosylate (BAY 43-9006 tosylate salt)
Sorafenib tosylate (BAY 43-9006 tosylate salt)
Formula: C₂₁H₁₆ClF₃N₄O₃·C₇H₈O₃S
MW: 637.03 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08235032
CAS Number: 475207-59-1
Order Now
Specification Test Results
Appearance Almost white to light brown powder
Infrared spectrum Conforms
HPLC ≥ 97.5%