Bedaquiline fumarate
Supplier: Thermo Fisher Scientific
Warning
Synonyms: (1R,2S)-1-(6-Bromo-2-methoxyquinolin-3-yl)-4-(dimethylamino)-2-(naphthalen-1-yl)-1-phenylbutan-2-ol fumarate
465910010 465912500
ACRO465910010EA 500 EUR
ACRO465910010 ACRO465912500
Bedaquiline fumarate
Bedaquiline fumarate
Formula: C₃₆H₃₅BrN₂O₆
MW: 671.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28167761
CAS Number: 845533-86-0
Order Now
Specification Test Results
Appearance White to almost white powder
Infrared spectrum Conforms
Water ≤ 1%
HPLC ≥ 98.5%
Fumaric acid 17.0 to 17.5%