Tetrabutylammonium bromide ≥98%
Supplier: VWR Chemicals
Warning
91243.150EA 23.8 EUR
91243.150 91243.230
Tetrabutylammonium bromide ≥98%
Tetrabutylammonium bromide
Formula: C₁₆H₃₆BrN
Melting Pt: 100…103 °C
Density: 1.148 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011633
CAS Number: 1643-19-2
EINECS: 216-699-2
Order Now
Specification Test Results
Assay Min. 98 %