C.T. agar dehydrated culture media
Supplier: HiMedia Laboratories
HIMEM1349-500GEA 126 EUR
HIMEM1349-500G
C.T. agar dehydrated culture media
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of myxobacteria.
  • Microbiological culture media
  • 500 g poly bottles