Agar No. 2 dehydrated culture media, Mueller Hinton
Supplier: HiMedia Laboratories
HIMEM1084-500GEA 103 EUR
HIMEM1084-500G
Agar No. 2 dehydrated culture media, Mueller Hinton
Media for Microbiology Dehydrated Media for Microbiology
For testing the susceptibility of common and rapidly growing bacteria using antimicrobial discs by the Bauer-Kirby method, manufactured to contain low levels of thymine, thymidine, calcium, and magnesium.
  • Microbiological culture media
  • 500 g poly bottles