Amidosulphonic acid ≥99.3% ACS

Supplier: Merck
Warning

SIAL383120-500GEA 203 EUR
SIAL383120-500G
Amidosulphonic acid ≥99.3% ACS
Amidosulphonic acid
Formula: H₃NO₃S
MW: 97,09 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 205 °C
Density: 2,15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011603
EINECS: 226-218-8
UN: 2967
ADR: 8,III
Merck Index: 12,09090
https://si.vwr.com/stibo/web/std.lang.all/75/87/2297587.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR