Face shields, V4 Series
Supplier: 3M
V4CK V4H
MMMAV4CKEA 0 EUR
MMMAV4CK MMMAV4H
Face shields, V4 Series
Face Shields
3M™ V4 series industrial Multisystem replacement face shields are available in multiple visor materials and color finishes.
  • Replacement face shields
  • Easy to install
  • Suitable for 3M™ PELTOR™ hearing protectors with headband
  • Thickness: 1,2 mm
  • Temperature range: – 40 to 100 °C
Ensure optimum comfort and protection from 3M™ earmuffs with headband with the 3M™ V40 Industrial Multisystem, V40G. These replacement face shields are available in a variety of visor materials and colours, and they are easy to install on 3M™ PELTOR™ hearing protectors with a headband.
Certifikace: Meets Grade F requirements for low energy impact protective shields. Marking class: 3-1.2 8 9 1F.