Reflective Materials, 9925 series, Scotchlite™
Supplier: 3M
7100117763 7100119093
MMMA7100117763EA 621 EUR
MMMA7100117763 MMMA7100119093
Reflective Materials, 9925 series, Scotchlite™
Tapes
3M™ Scotchlite™ Reflective material 9925 with high reflection values for high-visibility subject to higher stress, such as industrial and occupational garments that undergo industrial laundering.
  • Reflective woven fabric helps to enhance the visibility of the wearer at night or in low-light conditions
  • Very good washability in a disinfectant industrial laundering process at 75 °C with tumble drying at 90 °C or tunnel finishing at 155 °C
  • Can be used on delicate fabrics that can only be dry-cleaned
  • OEKO-TEX standard 100 certified
3M™ Scotchlite™ Reflective Material 9925 helps to enhance the visibility in low-light and night-time conditions. The material shines white when illuminated, even if the wearer is at the roadside. It meets various global high-visibility standards such as ISO 20471, and far exceeds the minimum reflections values required at the highest performance level. Reflective Material 9925 consists of high-precision glass beads bonded to a durable 100% polyester fabric. The product is washable in a home wash process at 60 °C (100 cycles according to ISO 6330-6N), and in an industrial wash process at 75 °C (75 cycles according to ISO 15797 table 4, procedure 2). It is also certified to the disinfectant industrial washing process of ISO 15797 table 1, procedure 2 (50 cycles at 75 °C). Dry cleaning is also possible (50 cycles as per ISO 3175-2). The material 9920 is OEKO-TEX standard 100 certified and contains no skin-irritating substances.