Ergonomic washable silicone mouse, GETT GCQ®
Supplier: Gett-Group
GETTKH24207EA 79.3 EUR
GETTKH24207
Ergonomic washable silicone mouse, GETT GCQ®
Computer Mice and Mouse Pads
This silicone mouse is a true all-rounder. It can be operated ergonomically and, thanks to the silicone surface, is perfect for application areas with high hygiene requirements.
  • Medical grade IP 68 mouse is perfect for applications with high hygiene requirements
  • Fully washable and disinfectable
  • Silicone cap to protect the USB connector from splashing water during cleaning
  • Ergonomic functionality with two mouse buttons as well as 3 scroll buttons
  • Laser sensor
  • Resolution: 1000 dpi
  • Button lifetime: >=2 mio. actuations
  • Engineered in Germany
  • 2-year warranty on the technical function
This silicone mouse has an ergonomic design and convinces with a pleasant tactile feeling at the highest degree of protection (IP 68). It can therefore be cleaned and disinfected all around. The mouse is a product of the GCQ series from GETT.
Certifikace: CE, FCC, REACH, RoHs, IP 68.
Balení: The mouse is delivered safely packed in brown cardboard box.