Acrylic acid 99% stabilised
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Prop-2-enoic acid
Stabilization: ∼200 ppm Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
L04280.22 L04280.36 L04280.0E L04280.0P
L04280.22EA 29.6 EUR
L04280.22 L04280.36 L04280.0E L04280.0P
Acrylic acid 99% stabilised
Acrylic acid
Formula: C₃H₄O₂
MW: 72.06 g/mol
Boiling Pt: 138…139 °C
Melting Pt: ∼ 13 °C
Density: 1.051
Flash Pt: 54 °C (130 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004367
CAS Number: 79-10-7
EINECS: 201-177-9
UN: 2218
ADR: 8 II
Merck Index: 14,130