6-Amino-3-fluoro-2-(trifluoromethoxy)phenol ≥80%, Technical Grade
Supplier: Apollo Scientific
Warning
PC502197-250MG PC502197-1G
APOSPC502197-250MGEA 21.9 EUR
APOSPC502197-250MG APOSPC502197-1G
6-Amino-3-fluoro-2-(trifluoromethoxy)phenol ≥80%, Technical Grade
6-Amino-3-fluoro-2-(trifluoromethoxy)phenol
Formula: C₇H₅F₄NO₂
MW: 211.11371 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28796316
CAS Number: 1806335-72-7