Methyl-2-bromo-1-methyl-1H-imidazole-5-carboxylate
Supplier: Apollo Scientific
Warning
OR59983-250MG OR59983-1G
APOSOR59983-250MGEA 132 EUR
APOSOR59983-250MG APOSOR59983-1G
Methyl-2-bromo-1-methyl-1H-imidazole-5-carboxylate
Methyl-2-bromo-1-methyl-1H-imidazole-5-carboxylate
Formula: C₆H₇BrN₂O₂
MW: 219.04 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD02179563
CAS Number: 120781-02-4
UN: 2811
ADR: 6 III