4-Fluoro-3-methylbenzylamine, Technical Grade
Supplier: Apollo Scientific
Synonyms:
(4-Fluoro-3-methylphenyl)methanamine
PC57806-5G
PC57806-25G
PC57806-100G
APOSPC57806-5GEA
21.9
EUR
APOSPC57806-5G
APOSPC57806-25G
APOSPC57806-100G
4-Fluoro-3-methylbenzylamine, Technical Grade
4-Fluoro-3-methylbenzylamine
Formula:
C₈H₁₀FN MW: 139.17 g/mol |
MDL Number:
MFCD01631490 CAS Number: 261951-68-2 UN: 2735 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...