2-Bromo-3-(trifluoromethyl)benzoic acid ≥95%
Supplier: Apollo Scientific
PC912791-1G PC912791-5G
APOSPC912791-1GEA 58 EUR
APOSPC912791-1G APOSPC912791-5G
2-Bromo-3-(trifluoromethyl)benzoic acid ≥95%
2-Bromo-3-(trifluoromethyl)benzoic acid
Formula: C₈H₄BrF₃O₂
MW: 269.02 g/mol
Melting Pt: 143…145 °C
MDL Number: MFCD08532493
CAS Number: 177420-63-2