Potassium permanganate concentrate for dilution to 0.002 mol (0.01N) in aqueous solution, ConvoL NORMADOSE® volumetric solution
Supplier: VWR Chemicals
Dose for preparation of 1 litre of solution.
91353.600EA 56.1 EUR
91353.600
Potassium permanganate concentrate for dilution to 0.002 mol (0.01N) in aqueous solution, ConvoL NORMADOSE® volumetric solution
Potassium permanganate
Formula: KMnO₄
MW: 158,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
Order Now
Specification Test Results
Molarity 0.00199 - 0.00201 mol/l (20°C)