BrdU Mouse Monoclonal Antibody [clone: A7] (ALEXA FLUOR® 680)

Supplier: Bioss
BOSSBSM-0917M-A680EA 1030 EUR
BOSSBSM-0917M-A680
BrdU Mouse Monoclonal Antibody [clone: A7] (ALEXA FLUOR® 680)
Antibodies
Bromodeoxyuridine is a synthetic nucleoside that is an analogue of thymidine. BrdU is incorporated into DNA during S-phase of a cell cycle. It is commonly used in the detection of proliferating cells in living tissues

Type: Primary
Antigen: Bromodeoxyuridine
Clonality: Monoclonal
Clone: A7
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Mouse
Isotype: IgG
Reactivity: Human; Mouse; Rat; Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR