Anti-Mycobacterium bovis Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BOSSBS-4622R-A680EA 840 EUR
BOSSBS-4622R-A680
Anti-Mycobacterium bovis Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Mycobacterium bovis.

Type: Primary
Antigen: Mycobacterium bovis
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria; Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR