Adenosine-5'-triphosphate disodium salt (ATP disodium salt) ≥99%
Supplier: Spectrum Chemical
SPCMAD125-1KGEA 0 EUR
SPCMAD125-1KG
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) ≥99%
Adenosine-5'-triphosphate disodium salt (ATP disodium salt)
Adenosine-5'-triphosphate Disodium Salt is a nucleoside triphosphate that cells use as a coenzyme. It is used by structural proteins and enzymes in several types of cellular processes that include motility and biosynthetic reactions.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula: C₁₀H₁₄N₅Na₂O₁₃P₃
MW: 551,14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00080339
EINECS: 213-579-1
Order Now
Specification Test Results
Assay (%) 99