Culti-Loops™ Morganella morganii ATCC™ 25830™
Supplier: Thermo Fisher Scientific
Culti-Loops™
OXOIR4604029EA 315 EUR
OXOIR4604029
Culti-Loops™ Morganella morganii ATCC™ 25830™
Microbiology Test Systems Microorganism Tests
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Balení: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.