Culti-Loops™ Proteus mirabilis 511-116
Supplier: Thermo Fisher Scientific
Culti-Loops™
OXOIR4608088EA 315 EUR
OXOIR4608088
Culti-Loops™ Proteus mirabilis 511-116
Microbiology Test Systems Microorganism Tests
Confidently carry out testing of media, reagents, and identification and antimicrobial susceptibility kits and evaluate the bacteriological procedures with Culti-Loops™ ready-to-use QC organisms. Proteus mirabilis 511-116 strain produces extended-spectrum β-lactamase (ESBL). This strain is recommended to be used as a positive control strain with Spectra™ ESBL Medium following the HAI surveillance programs.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Balení: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Upozornení: For in vitro diagnostic use.