Culti-Loops™ Bacillus cereus ATCC™ 14579™
Supplier: Thermo Fisher Scientific
Culti-Loops™
OXOIR4601217EA 315 EUR
OXOIR4601217
Culti-Loops™ Bacillus cereus ATCC™ 14579™
Microbiology Test Systems Microorganism Tests
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Balení: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.