Printer Ribbons, M710 R6800 Series
Supplier: Brady
BRDY173185EA 60.67 EUR
BRDY173185
Printer Ribbons, M710 R6800 Series
Printer Ribbons
One-color ribbon for printing a single color onto a label.
  • R6800 series white resin ribbons offer excellent solvent, scratch, and smudge resistance when printed on the appropriate Brady® label material
  • Recommended for printing on dark-colored, heat shrinkable tubing materials, such as Brady® black B-345 PermaSleeve® high temperature wire markers
  • Self-contained M710 series ribbon cartridges insert straight into the printer with ease
M710 Series snap-in print ribbons enable 5 second ribbon supply changes. No calibrating, no threading, no waste, no hassles, and the ink always faces the right direction.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.